N1-[2-(3,4-Dimethylphenoxy)ethyl]-N4,N4-diethyl-1,4-benzenediamine
Supplier: Matrix Scientific
032240-500MG
101833-304EA
306.15
USD
101833-304
N1-[2-(3,4-Dimethylphenoxy)ethyl]-N4,N4-diethyl-1,4-benzenediamine
N1-[2-(3,4-Dimethylphenoxy)ethyl]-N4,N4-diethyl-1,4-benzenediamine
Hazard: Irritant
Formula:
C₂₀H₂₈N₂O |
MDL Number:
MFCD10688240 CAS Number: 1040680-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...