3-(Heptyloxy)-N-[2-(2-phenoxyethoxy)benzyl]aniline
Supplier: Matrix Scientific
032238-500MG
101833-300EA
306.15
USD
101833-300
3-(Heptyloxy)-N-[2-(2-phenoxyethoxy)benzyl]aniline
3-(Heptyloxy)-N-[2-(2-phenoxyethoxy)benzyl]aniline
Hazard: Irritant
Formula:
C₂₈H₃₅NO₃ |
MDL Number:
MFCD10688238 CAS Number: 1040680-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...