N-[2-(4-Ethylphenoxy)propyl]-3-(heptyloxy)aniline

Supplier: Matrix Scientific

032237-500MG
101833-298EA 306.15 USD
101833-298
N-[2-(4-Ethylphenoxy)propyl]-3-(heptyloxy)aniline
N-[2-(4-Ethylphenoxy)propyl]-3-(heptyloxy)aniline

Hazard: Irritant

Formula: C₂₄H₃₅NO₂
MDL Number: MFCD10688237
CAS Number: 1040680-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR