N-[2-(2-Chlorophenoxy)propyl]-4-(2-cyclohexylethoxy)aniline
Supplier: Matrix Scientific
032225-500MG
101833-274EA
306.15
USD
101833-274
N-[2-(2-Chlorophenoxy)propyl]-4-(2-cyclohexylethoxy)aniline
N-[2-(2-Chlorophenoxy)propyl]-4-(2-cyclohexylethoxy)aniline
Hazard: Irritant
Formula:
C₂₃H₃₀ClNO₂ |
MDL Number:
MFCD10688225 CAS Number: 1040693-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...