N-[4-(2-Methoxyethoxy)benzyl]-3,4-dimethylaniline

Supplier: Matrix Scientific

032173-500MG
101833-170EA 306.15 USD
101833-170
N-[4-(2-Methoxyethoxy)benzyl]-3,4-dimethylaniline
N-[4-(2-Methoxyethoxy)benzyl]-3,4-dimethylaniline

Hazard: Irritant

Formula: C₁₈H₂₃NO₂
MDL Number: MFCD10688179
CAS Number: 1040690-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR