N-[2-(2-Ethoxyethoxy)benzyl]-2-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032139-500MG
101833-102EA 306.15 USD
101833-102
N-[2-(2-Ethoxyethoxy)benzyl]-2-(3-phenylpropoxy)aniline
N-[2-(2-Ethoxyethoxy)benzyl]-2-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₂₆H₃₁NO₃
MDL Number: MFCD10688145
CAS Number: 1040688-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR