N-(2-Isobutoxybenzyl)-2-methyl-2-propanamine

Supplier: Matrix Scientific

031590-500MG
101832-006EA 306.15 USD
101832-006
N-(2-Isobutoxybenzyl)-2-methyl-2-propanamine
N-(2-Isobutoxybenzyl)-2-methyl-2-propanamine

Hazard: Irritant

Formula: C₁₅H₂₅NO
MDL Number: MFCD10687657
CAS Number: 1040689-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR