N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-4-ethoxyaniline

Supplier: Matrix Scientific

031425-500MG
101831-676EA 306.15 USD
101831-676
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-4-ethoxyaniline
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-4-ethoxyaniline

Hazard: Irritant

Formula: C₁₈H₂₂ClNO₂
MDL Number: MFCD10687537
CAS Number: 1040686-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR