N,2,6-Trimethylaniline

Supplier: Matrix Scientific

031419-500MG
101831-664EA 221.54 USD
101831-664
N,2,6-Trimethylaniline
N,2,6-Trimethylaniline

Hazard: Irritant

Formula: C₉H₁₃N
MDL Number: MFCD00613418
CAS Number: 767-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR