N-(4-Butoxybenzyl)(3-pyridinyl)methanamine

Supplier: Matrix Scientific

031392-500MG
101831-610EA 306.15 USD
101831-610
N-(4-Butoxybenzyl)(3-pyridinyl)methanamine
N-(4-Butoxybenzyl)(3-pyridinyl)methanamine

Hazard: Irritant

Formula: C₁₇H₂₂N₂O
MDL Number: MFCD10687514
CAS Number: 1040685-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR