N-[4-(2-Ethoxyethoxy)benzyl]-3-isopropoxyaniline

Supplier: Matrix Scientific

031313-500MG
101831-452EA 306.15 USD
101831-452
N-[4-(2-Ethoxyethoxy)benzyl]-3-isopropoxyaniline
N-[4-(2-Ethoxyethoxy)benzyl]-3-isopropoxyaniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₃
MDL Number: MFCD10687444
CAS Number: 1040684-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR