N-(4-Ethoxybenzyl)-3-(phenethyloxy)aniline

Supplier: Matrix Scientific

031277-500MG
101831-380EA 306.15 USD
101831-380
N-(4-Ethoxybenzyl)-3-(phenethyloxy)aniline
N-(4-Ethoxybenzyl)-3-(phenethyloxy)aniline

Hazard: Irritant

Formula: C₂₃H₂₅NO₂
MW: 347.46 g/mol
MDL Number: MFCD10687408
CAS Number: 1040682-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR