3-Bromo-2-chloro-5-(trifluoromethyl)pyridine ≥98% for synthesis

Supplier: Matrix Scientific

031271-1G
101831-368EA 15.38 USD
101831-368
3-Bromo-2-chloro-5-(trifluoromethyl)pyridine ≥98% for synthesis
3-Bromo-2-chloro-5-(trifluoromethyl)pyridine

Hazard code(s): Irritant

Formula: C₆H₂BrClF₃N
MDL Number: MFCD09878432
CAS Number: 71701-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR