4-Bromo-1'-methyl-1H,1'H-3,4'-bipyrazole

Supplier: Matrix Scientific

030914-500MG
101831-090EA 500 USD
101831-090
4-Bromo-1'-methyl-1H,1'H-3,4'-bipyrazole
4-Bromo-1'-methyl-1H,1'H-3,4'-bipyrazole

Hazard: Irritant

Formula: C₇H₇BrN₄
MDL Number: MFCD04970825
CAS Number: 925146-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR