[(9-Methyl-9H-[1,2,4]triazolo[4,3-a]benzimidazol-3-yl)thio]acetic acid

Supplier: Matrix Scientific

030703-500MG
101830-664EA 387.69 USD
101830-664
[(9-Methyl-9H-[1,2,4]triazolo[4,3-a]benzimidazol-3-yl)thio]acetic acid
[(9-Methyl-9H-[1,2,4]triazolo[4,3-a]benzimidazol-3-yl)thio]acetic acid

Hazard: Irritant

Formula: C₁₁H₁₀N₄O₂S
MDL Number: MFCD03767538
CAS Number: 681838-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR