5-(2-Methoxyphenyl)-4-methyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030401-500MG
101830-064EA 146.15 USD
101830-064
5-(2-Methoxyphenyl)-4-methyl-4H-1,2,4-triazole-3-thiol
5-(2-Methoxyphenyl)-4-methyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₀H₁₁N₃OS
MDL Number: MFCD02091580
CAS Number: 58064-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR