Staticmaster® Duostat Positioner, NRD
Supplier: NRD Staticmaster
BF2
10183-836EA 249.25 USD
10183-836
Staticmaster® Duostat Positioner, NRD
Ionizers
The Staticmaster® BF2-1000 Duostat Positioner is the ideal tool for removing static charge from filter media and fine powders.
  • Increases weighing accuracy
  • Useful for filter media and fine powders
  • Removes static charge prior to placement in weigh chamber
The Duostat Positioner, when combined with two (2), 2U500 Ionizing Cartridges (sold separately) removes static charge from filter media and fine powders prior to placing them into the weigh chamber. Utilizing the BF2-1000 system results in increased weighing accuracy and efficient, reliable measurement.