Staticmaster® 2U500 Grounding Kit, NRD
Supplier: NRD Staticmaster
5060
10183-826EA 89.96 USD
10183-826
Staticmaster® 2U500 Grounding Kit, NRD
Ionizers
The 2U500 Grounding Kit significantly enhances the static eliminating power of the 2U500 Ionizing Cartridge.
  • Enhances static eliminating power
  • Increases performance of static eliminators
Add the Grounding Kit to your in-balance ionizer to increase the performance of the static eliminator. Adapter is made of ABS/PC. Tinned copper ground wire is 4'.