Y1451 Thermal Liners, Wells Lamont
Supplier: Wells Lamont
Y1451
10183-812PK 78.35 USD
10183-812
Y1451 Thermal Liners, Wells Lamont
Gloves
The white thermal liners combine comfort with thermal protection by conforming to the hand.
  • Gloves work alone or as liners
  • Gloves wick away moisture
  • More hygienic than cotton liners
  • Thermal
  • Gloves conform to hand
  • Hollow core polyester/Lycra® construction
Ordering information: Glove liners are sold by the dozen pairs.