N,N-Diethyl-4-isothiocyanatobenzenesulfonamide

Supplier: Matrix Scientific

030259-500MG
101829-784EA 336.92 USD
101829-784
N,N-Diethyl-4-isothiocyanatobenzenesulfonamide
N,N-Diethyl-4-isothiocyanatobenzenesulfonamide

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂S₂
MDL Number: MFCD09971961
CAS Number: 99841-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR