6-Fluoro-1-[(4-isothiocyanatophenyl)sulfonyl]-2-methyl-1,2,3,4-tetrahydroquinoline

Supplier: Matrix Scientific

030256-500MG
101829-778EA 193.85 USD
101829-778
6-Fluoro-1-[(4-isothiocyanatophenyl)sulfonyl]-2-methyl-1,2,3,4-tetrahydroquinoline
6-Fluoro-1-[(4-isothiocyanatophenyl)sulfonyl]-2-methyl-1,2,3,4-tetrahydroquinoline

Hazard: Irritant

Formula: C₁₇H₁₅FN₂O₂S₂
MDL Number: MFCD09971958
CAS Number: 956576-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR