3-Bromophenyl isocyanate

Supplier: Matrix Scientific

030188-500MG
101829-656EA 146.15 USD
101829-656
3-Bromophenyl isocyanate
3-Bromophenyl isocyanate
Formula: C₇H₄BrNO
Boiling Pt: 220 °C (1013 hPa)
Density: 1.586 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002014
CAS Number: 23138-55-8
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR