4-Fluorobenzenesulfonohydrazide

Supplier: Matrix Scientific

029907-500MG
101829-090EA 193.85 USD
101829-090
4-Fluorobenzenesulfonohydrazide
4-Fluorobenzenesulfonohydrazide

Hazard: Irritant

Formula: C₆H₇FN₂O₂S
MW: 190.2 g/mol
MDL Number: MFCD00051748
CAS Number: 2266-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR