2-(2,4-Dimethylphenoxy)propanohydrazide

Supplier: Matrix Scientific

029900-500MG
101829-076EA 146.15 USD
101829-076
2-(2,4-Dimethylphenoxy)propanohydrazide
2-(2,4-Dimethylphenoxy)propanohydrazide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MW: 208.26 g/mol
MDL Number: MFCD03423276
CAS Number: 125096-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR