2-[(6-Bromo-2-naphthyl)oxy]acetohydrazide

Supplier: Matrix Scientific

029877-500MG
101829-034EA 290.77 USD
101829-034
2-[(6-Bromo-2-naphthyl)oxy]acetohydrazide
2-[(6-Bromo-2-naphthyl)oxy]acetohydrazide

Hazard: Irritant

Formula: C₁₂H₁₁BrN₂O₂
MW: 295.14 g/mol
MDL Number: MFCD00245485
CAS Number: 438219-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR