2-Amino-6-tert-butyl-N-(2-methylphenyl)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
Supplier: Matrix Scientific
029114-500MG
101827-502EA
193.85
USD
101827-502
2-Amino-6-tert-butyl-N-(2-methylphenyl)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
2-Amino-6-tert-butyl-N-(2-methylphenyl)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
CAS: 433251-65-1
MDL Number: MFCD03145441
Molecular Formula: C20H26N2OS
Molecular Weight: 342,51
Hazard: Irritant
Formula:
C₂₀H₂₆N₂OS |
MDL Number:
MFCD03145441 CAS Number: 433251-65-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...