2-Amino-N-(4-methylphenyl)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
Supplier: Matrix Scientific
029099-500MG
101827-472EA
193.85
USD
101827-472
2-Amino-N-(4-methylphenyl)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
2-Amino-N-(4-methylphenyl)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
CAS: 83822-35-9
MDL Number: MFCD00421038
Molecular Formula: C16H18N2OS
Molecular Weight: 286,4
Hazard: Irritant
Formula:
C₁₆H₁₈N₂OS |
MDL Number:
MFCD00421038 CAS Number: 83822-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...