Ethyl-2-amino-4-(3,4-dimethylphenyl)thiophene-3-carboxylate

Supplier: Matrix Scientific

028947-500MG
101827-168EA 193.85 USD
101827-168
Ethyl-2-amino-4-(3,4-dimethylphenyl)thiophene-3-carboxylate
Ethyl-2-amino-4-(3,4-dimethylphenyl)thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₇NO₂S
MDL Number: MFCD01917263
CAS Number: 307511-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR