4-Methoxy-3-(thiomorpholin-4-ylmethyl)benzaldehyde

Supplier: Matrix Scientific

028406-500MG
101826-072EA 193.85 USD
101826-072
4-Methoxy-3-(thiomorpholin-4-ylmethyl)benzaldehyde
4-Methoxy-3-(thiomorpholin-4-ylmethyl)benzaldehyde

Hazard: Irritant

Formula: C₁₃H₁₇NO₂S
MDL Number: MFCD01114960
CAS Number: 438531-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR