6-Methyl-2-(4-methylphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027884-500MG
101825-044EA 146.15 USD
101825-044
6-Methyl-2-(4-methylphenyl)quinoline-4-carboxylic acid
6-Methyl-2-(4-methylphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₅NO₂
MW: 277.33 g/mol
MDL Number: MFCD01825280
CAS Number: 18060-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR