5-(Azidomethyl)quinolin-8-ol

Supplier: Matrix Scientific

027789-500MG
101824-856EA 387.69 USD
101824-856
5-(Azidomethyl)quinolin-8-ol
5-(Azidomethyl)quinolin-8-ol

Hazard: Irritant

Formula: C₁₀H₈N₄O
MW: 200.2 g/mol
MDL Number: MFCD09759044
CAS Number: 1005143-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR