N-Methyl-N-[(5-phenyl-1H-pyrazol-4-yl)methyl]amine

Supplier: Matrix Scientific

027637-500MG
101824-572EA 387.69 USD
101824-572
N-Methyl-N-[(5-phenyl-1H-pyrazol-4-yl)methyl]amine
N-Methyl-N-[(5-phenyl-1H-pyrazol-4-yl)methyl]amine

Hazard: Irritant

Formula: C₁₁H₁₃N₃
MDL Number: MFCD08691429
CAS Number: 1007521-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR