[(6-Methyl-1H-benzimidazol-2-yl)methoxy]acetic acid

Supplier: Matrix Scientific

027613-500MG
101824-524EA 290.77 USD
101824-524
[(6-Methyl-1H-benzimidazol-2-yl)methoxy]acetic acid
[(6-Methyl-1H-benzimidazol-2-yl)methoxy]acetic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MW: 220.23 g/mol
MDL Number: MFCD07643236
CAS Number: 876716-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR