(2E)-1-Methyl-2-(nitromethylene)pyrrolidine

Supplier: Matrix Scientific

027588-500MG
101824-476EA 387.69 USD
101824-476
(2E)-1-Methyl-2-(nitromethylene)pyrrolidine
(2E)-1-Methyl-2-(nitromethylene)pyrrolidine

Hazard: Irritant

Formula: C₆H₁₀N₂O₂
MW: 142.16 g/mol
MDL Number: MFCD00222734
CAS Number: 94662-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR