4-Bromo-3-methyl-5-nitro-1H-pyrazole

Supplier: Matrix Scientific

027403-500MG
101824-120EA 692.31 USD
101824-120
4-Bromo-3-methyl-5-nitro-1H-pyrazole
4-Bromo-3-methyl-5-nitro-1H-pyrazole

Hazard: Irritant

Formula: C₄H₄BrN₃O₂
MDL Number: MFCD01459884
CAS Number: 70951-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR