N,N-Dimethylglycine ≥98% for synthesis
Supplier: Matrix Scientific
Synonyms:
2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid
027373-25G
027373-100G
101824-050EA
29.23
USD
101824-050
101824-052
N,N-Dimethylglycine ≥98% for synthesis
N,N-Dimethylglycine
Hazard code(s): Irritant
Formula:
C₄H₉NO₂ |
MDL Number:
MFCD00004283 CAS Number: 1118-68-9 EINECS: 214-267-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...