2-Phenylbenzoic acid

Supplier: Matrix Scientific

Synonyms: Biphenyl-2-carboxylic acid, 2-Biphenylcarboxylic acid, [1,1'-Biphenyl]-2-carboxylic acid

027210-1G
101823-736EA 15.38 USD
101823-736
2-Phenylbenzoic acid
2-Phenylbenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₀O₂
MDL Number: MFCD00002463
CAS Number: 947-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR