3-{[4-(Diethylamino)benzyl]amino}propanamide

Supplier: Matrix Scientific

026805-500MG
101822-906EA 306.15 USD
101822-906
3-{[4-(Diethylamino)benzyl]amino}propanamide
3-{[4-(Diethylamino)benzyl]amino}propanamide

Hazard: Irritant

Formula: C₁₄H₂₃N₃O
MDL Number: MFCD10687266
CAS Number: 1040688-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR