Thiophene-2-carboxylic acid (4-amino-2-methoxyphenyl)amide

Supplier: Matrix Scientific

026643-500MG
101822-614EA 306.15 USD
101822-614
Thiophene-2-carboxylic acid (4-amino-2-methoxyphenyl)amide
Thiophene-2-carboxylic acid (4-amino-2-methoxyphenyl)amide

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂S
MDL Number: MFCD02258051
CAS Number: 110506-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR