5-Methoxynicotinaldehyde

Supplier: Matrix Scientific

026164-250MG
101821-732EA 20 USD
101821-732
5-Methoxynicotinaldehyde
5-Methoxynicotinaldehyde

Hazard: Irritant

Formula: C7H7NO2
MDL Number: MFCD07437946
CAS Number: 113118-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR