N-(5-Amino-2-methoxyphenyl)-3-isobutoxybenzamide
Supplier: Matrix Scientific
025894-500MG
101821-192EA
306.15
USD
101821-192
N-(5-Amino-2-methoxyphenyl)-3-isobutoxybenzamide
N-(5-Amino-2-methoxyphenyl)-3-isobutoxybenzamide
CAS: 1020055-24-6
MDL Number: MFCD09997490
Molecular Formula: C18H22N2O3
Molecular Weight: 314,39
Hazard: Irritant
Formula:
C₁₈H₂₂N₂O₃ |
MDL Number:
MFCD09997490 CAS Number: 1020055-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...