N-(5-Amino-2-methoxyphenyl)-2-[2-(sec-butyl)phenoxy]propanamide
Supplier: Matrix Scientific
025891-500MG
101821-186EA
306.15
USD
101821-186
N-(5-Amino-2-methoxyphenyl)-2-[2-(sec-butyl)phenoxy]propanamide
N-(5-Amino-2-methoxyphenyl)-2-[2-(sec-butyl)phenoxy]propanamide
CAS: 1020055-16-6
MDL Number: MFCD09997488
Molecular Formula: C20H26N2O3
Molecular Weight: 342,44
Hazard: Irritant
Formula:
C₂₀H₂₆N₂O₃ |
MDL Number:
MFCD09997488 CAS Number: 1020055-16-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...