N-(3-Amino-2-methylphenyl)-2-furamide

Supplier: Matrix Scientific

025325-500MG
101820-028EA 306.15 USD
101820-028
N-(3-Amino-2-methylphenyl)-2-furamide
N-(3-Amino-2-methylphenyl)-2-furamide

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD01974785
CAS Number: 900636-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR