N-(4-Aminophenyl)-2-phenylacetamide

Supplier: Matrix Scientific

025265-500MG
101819-908EA 306.15 USD
101819-908
N-(4-Aminophenyl)-2-phenylacetamide
N-(4-Aminophenyl)-2-phenylacetamide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O
MDL Number: MFCD03906887
CAS Number: 346583-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR