N-(3-Aminophenyl)-4-(2-methoxyethoxy)benzamide

Supplier: Matrix Scientific

025090-500MG
101819-558EA 306.15 USD
101819-558
N-(3-Aminophenyl)-4-(2-methoxyethoxy)benzamide
N-(3-Aminophenyl)-4-(2-methoxyethoxy)benzamide

Hazard: Irritant

Formula: C₁₆H₁₈N₂O₃
MDL Number: MFCD09997109
CAS Number: 1020722-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR