N-(3-Aminophenyl)-2-(2-methylphenoxy)butanamide

Supplier: Matrix Scientific

025047-500MG
101819-472EA 306.15 USD
101819-472
N-(3-Aminophenyl)-2-(2-methylphenoxy)butanamide
N-(3-Aminophenyl)-2-(2-methylphenoxy)butanamide

Hazard: Irritant

Formula: C₁₇H₂₀N₂O₂
MW: 284.36 g/mol
MDL Number: MFCD09997087
CAS Number: 1020722-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR