1-[(2-Methyl-4-methylsulfonyl-6-nitro)phenyl]piperazine

Supplier: Matrix Scientific

024867-500MG
101819-110EA 364.62 USD
101819-110
1-[(2-Methyl-4-methylsulfonyl-6-nitro)phenyl]piperazine
1-[(2-Methyl-4-methylsulfonyl-6-nitro)phenyl]piperazine

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₄S
Melting Pt: 214…217 °C
MDL Number: MFCD09264541
CAS Number: 1000018-29-0

Order Now

Specification Test Results

Melting point 166 - 168


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR