[1-(3-Nitropyridin-2-yl)piperazin-4-yl]ethanol hydrochloride

Supplier: Matrix Scientific

024852-500MG
101819-082EA 253.85 USD
101819-082
[1-(3-Nitropyridin-2-yl)piperazin-4-yl]ethanol hydrochloride
[1-(3-Nitropyridin-2-yl)piperazin-4-yl]ethanol hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₇ClN₄O₃
MDL Number: MFCD09997078
CAS Number: 1185299-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR