4-Acetylbenzenesulfonamide ≥97%

Supplier: Matrix Scientific

024827-1G
101819-006EA 58.46 USD
101819-006
4-Acetylbenzenesulfonamide ≥97%
4-Acetylbenzenesulfonamide

Hazard: Irritant

Formula: C₈H₉NO₃S
MDL Number: MFCD00792524
CAS Number: 1565-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR