1-[(2-Chloro-4-methylsulfonyl)phenyl]homopiperazine hydrochloride

Supplier: Matrix Scientific

024522-250MG
101818-248EA 224.62 USD
101818-248
1-[(2-Chloro-4-methylsulfonyl)phenyl]homopiperazine hydrochloride
1-[(2-Chloro-4-methylsulfonyl)phenyl]homopiperazine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈Cl₂N₂O₂S
MDL Number: MFCD09038313
CAS Number: 1170490-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR